Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN3 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB270743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270743 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270743 Supplier Novus Biologicals Supplier No. NBP244286R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAN3 Polyclonal antibody specifically detects PAN3 in Human samples. It is validated for Western Blot

Specifications

Antigen PAN3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias hPan3, PAB-dependent poly(A)-specific ribonuclease subunit 3, PABP1-dependent poly A-specific ribonuclease subunit PAN3, PABP-dependent poly(A) nuclease 3, PAN3 poly(A) specific ribonuclease subunit homolog (S. cerevisiae), TS
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residue 175 to 225 of human PAB-dependent poly using the numbering given in entry Q58A45.3 (GeneID 255967).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 255967
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.