Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAN3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571860
Description
PAN3 is functions in cytoplasmic mRNA decay. It is as part of the Pan nuclease complex, recruits polyadenylate-binding protein which in turn stimulates PAN2 nuclease activity.
Specifications
PAN3 | |
Polyclonal | |
Unconjugated | |
PAN3 | |
PAB-dependent poly(A)-specific ribonuclease subunit PAN3 {ECO:0000255/HAMAP-Rule:MF_03181}; PAB1P-dependent poly(A)-nuclease {ECO:0000255/HAMAP-Rule:MF_03181}; PAN deadenylation complex subunit 3 {ECO:0000255/HAMAP-Rule:MF_03181}; Pan3 | |
Rabbit | |
Protein A | |
RUO | |
255967, 72587 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.97 mg/mL | |
PBS with 0.09% sodium azide | |
Q58A45, Q640Q5 | |
PAN3 | |
KLH conjugated synthetic peptide between 727-755 amino acids from the C-terminal region of mouse Pan3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction