Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Amylase Alpha Antibody - BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
$207.00 - $499.50
Specifications
Antigen | Pancreatic Amylase Alpha |
---|---|
Concentration | 5.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017767
![]() |
Novus Biologicals
NB100663880.1ML |
0.1 mL |
Each for $207.00
|
|
|||||
NB10066388
![]() |
Novus Biologicals
NB10066388 |
0.5 mL |
Each for $499.50
|
|
|||||
Description
Pancreatic Amylase Alpha Polyclonal specifically detects Pancreatic Amylase Alpha in Human, Porcine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Pancreatic Amylase Alpha | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
Polyclonal | |
Purified | |
RUO | |
Phosphate buffered saline with 0.09% Sodium Azide | |
AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase | |
AMY2A | |
IgG | |
IgG purified | |
Detects a major band of approximately 56 kDa in porcine amylase samples under reducing conditions. |
5.0 mg/mL | |
Immunofluorescence, Immunocytochemistry, Western Blot, ELISA | |
Unconjugated | |
Sheep | |
Human, Pig | |
P00690 | |
279 | |
Porcine pancreatic alpha-amylase | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title