Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Amylase Alpha Antibody (OTI6D4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147659
Description
Pancreatic Amylase Alpha Monoclonal specifically detects Pancreatic Amylase Alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pancreatic Amylase Alpha | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 | |
AMY2A | |
Recombinant protein expressed in E.coli corresponding to amino acids 300-511 of human AMY2A | |
Affinity Purified | |
RUO | |
279 | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI6D4 | |
Unconjugated | |
AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase | |
Mouse | |
57.7 kDa | |
0.1 mL | |
Primary | |
This antibody is specific for Homo sapiens amylase, alpha 2A (pancreatic) (AMY2A). | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction