Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Amylase Alpha Antibody (OTI6D4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273238PCP
Description
Pancreatic Amylase Alpha Monoclonal antibody specifically detects Pancreatic Amylase Alpha in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Pancreatic Amylase Alpha | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
279 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI6D4 | |
Western Blot, Immunohistochemistry | |
AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase | |
Recombinant protein expressed in E.coli corresponding to amino acids 300-511 of human AMY2A | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction