Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Amylase Beta Antibody (OTI4B5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273240H
Description
Pancreatic Amylase Beta Monoclonal specifically detects Pancreatic Amylase Beta in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Pancreatic Amylase Beta | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
AMY2B | |
Full length recombinant protein of human Pancreatic Amylase Beta (NP_066188) produced in HEK293T cells. | |
0.1 mL | |
280.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4B5 | |
Western Blot, Immunohistochemistry | |
alpha-amylase 2B, AMY2, AMY2B amylase, alpha 2B (pancreatic) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction