Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic Amylase Beta Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00

Specifications

Antigen Pancreatic Amylase Beta
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095498
SDP
View Documents
Novus Biologicals
NBP30350020UL
20 μL
Each of 1 for $206.00
Only null left
Add to Cart
 
Description

Description

Pancreatic Amylase Beta Polyclonal specifically detects Pancreatic Amylase Beta in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Pancreatic Amylase Beta
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
280
IgG
Affinity purified
Immunofluorescence
Unconjugated
Rabbit
Human
alpha-amylase 2B, AMY2, AMY2B amylase, alpha 2B (pancreatic)
Recombinant fusion protein containing a sequence corresponding to amino acids 16-320 of human Pancreatic Amylase Beta (NP_066188.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.