Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pancreatic Amylase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117115 Supplier Invitrogen™ Supplier No. PA5117115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse intestine, Rat intestine Immunogen sequence: QYSPNTQQGR TSIVHLFEWR WVDIALECER YLAPKGFGGV QVSPPNENVA IYNPFRPWWE RYQPVSYKLC TRSGNEDEFR NMVTRCNNVG VRIYVDAVIN HMCGNAVSAG TSSTCGSYFN PGSRDFPAVP YSGWDFNDGK CKTGSGDIEN YNDATQVRDC RLTGLLDLAL EKDYVRSKIA EYMNHLIDIG VAGFRLDASK HMWPGDIKAI LDKLHNLNSN WFPAGSKPFI YQEVIDLGGE PIKSSDYFGN GRVTEFKYGA KLGTVIRKWN GEKMSYLKNW GEGWGFVPSD RALVFVDNHD NQRGHGAGGA SILTFWDARL YKMAVGFMLA HPYGFTRVMS SYRWPRQFQN GNDVNDWVGP PNNNGVIKEV TINPDTTCGN DWVCEHRWRQ IRNMVIFRNV VDGQPFTNWY DNGSNQVAFG RGNRGFIVFN NDDWSFSLTL QTGLPAGTYC DVISGDKING NCTGIKIYVS DDGKAHFSIS NSAEDPFIAI HAESKL.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch andglycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pancreatic Amylase
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AMY2B
Gene Accession No. P04746, P19961
Gene Alias 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Symbols AMY2A, Amy2a1, Amy2a4, AMY2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 16-511 of human AMY2A (NP_0006901).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100043207, 100043684, 279, 280, 545562
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.