Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Lipase Antibody (114), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290230UV
Description
Pancreatic Lipase Monoclonal specifically detects Pancreatic Lipase in Human samples. It is validated for ELISA.Specifications
Pancreatic Lipase | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
5406 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
114 | |
ELISA | |
EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Pancreatic Lipase (P16233; Met1-Cys465). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction