Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Lipase Rabbit anti-Human, Clone: 120, Novus Biologicals™

Rabbit Monoclonal Antibody
$345.19 - $554.59
Specifications
| Antigen | Pancreatic Lipase |
|---|---|
| Clone | 120 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
Pancreatic Lipase Monoclonal specifically detects Pancreatic Lipase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Pancreatic Lipase | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 5406 | |
| IgG | |
| Protein A purified |
| 120 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase | |
| Recombinant Human Pancreatic Lipase Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title