Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Polypeptide/PP Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 16 publications
$231.00 - $562.50
Specifications
Antigen | Pancreatic Polypeptide/PP |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Pancreatic Polypeptide/PP Polyclonal specifically detects Pancreatic Polypeptide/PP in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Peptide ELISA.Specifications
Pancreatic Polypeptide/PP | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, ELISA | |
Unconjugated | |
RUO | |
Human, Mouse, Monkey | |
P01298 | |
5539 | |
Peptide with sequence C-TRPRYGKRHKEDT corresponding to internal region according to NP_002713.1. | |
Primary |
0.5 mg/mL | |
Polyclonal | |
Goat | |
Stem Cell Markers | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone | |
PPY | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title