Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic Polypeptide Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. AB939I25UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB939-I-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB939-I-25UL Supplier MilliporeSigma Supplier No. AB939I25UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Pancreatic Polypeptide Antibody, Cat. No. AB939-I, is a highly specific rabbit polyclonal antibody that targets Pancreatic polypeptide and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pancreatic Polypeptide
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Pancreatic hormone;PP;Obinepitide
Gene Symbols PPY;PNP
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 36 amino acids the internal region of human pancreatic polypeptide.
Molecular Weight of Antigen ∽11 and ∽20 kDa observed for isoforms 1 and 2; 10.45 and 18.79 kDa calculated for isoforms 1 and 2. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects both isoforms of human pancreatic polypeptide. It targets an epitope within 36 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.