Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Panendothelial Cell Antigen Antibody (MECA-32), Allophycocyanin/Cy7, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10077668APCCY7
Description
Panendothelial Cell Antigen Monoclonal specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| Panendothelial Cell Antigen | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Plvap | |
| Mouse lymph node stromal cells. | |
| 0.1 mL | |
| Primary | |
| Pan endothelial cells except for CNS & muscle capillaries. | |
| Store at 4°C in the dark. Do not freeze. | 
| Flow Cytometry | |
| MECA-32 | |
| Flow Cytometry | |
| FELS, Fenestrated endothelial-linked structure protein, gp68, MECA32, MECA-32, Panendothelial Cell Antigen, plasmalemma vesicle associated protein, Plasmalemma Vesicle Protein 1, PV-1 protein | |
| Rat | |
| Protein G purified | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 84094.0 | |
| Human, Mouse | |
| IgG2a κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction