Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), PE/Atto594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10077668PEATT594
Description
Panendothelial Cell Antigen Monoclonal specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Panendothelial Cell Antigen | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
Plvap | |
Mouse lymph node stromal cells. | |
0.1 mL | |
Primary | |
Pan endothelial cells except for CNS & muscle capillaries. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MECA-32 | |
Flow Cytometry | |
FELS, Fenestrated endothelial-linked structure protein, gp68, MECA32, MECA-32, Panendothelial Cell Antigen, plasmalemma vesicle associated protein, Plasmalemma Vesicle Protein 1, PV-1 protein | |
Rat | |
Protein G purified | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
84094.0 | |
Human, Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction