Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PANK1/2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 124661AP150UL

Description
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is expressed most abundantly in the liver.Specifications
PANK1/2/3 | |
Polyclonal | |
Unconjugated | |
PANK1 | |
hPanK3, PANK1/2/3, PANK3, pantothenate kinase 3, Pantothenic acid kinase 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
211347, 294088, 296167, 360511, 53354, 74450, 75735, 79646, 80025 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K4K6, Q8R2W9, Q8TE04, Q9BZ23, Q9H999 | |
Pank1, Pank2, Pank3 | |
PANK1/2/3 Fusion Protein Ag3140 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction