Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PANK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116411
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116411 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116411 Supplier Invitrogen™ Supplier No. PA5116411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PANK1 Polyclonal Antibody for Western Blot

PANK1 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PANK1
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PANK1
Gene Accession No. Q8K4K6, Q8TE04
Gene Alias hPanK; hPanK1; mPank; Pank; PANK1; Pank1b; Pantothenate kinase 1; pantothenic acid kinase 1
Gene Symbols PANK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 75-105 amino acids from the N-terminal region of human PANK1.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53354, 75735
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.