Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PANK2 Antibody (OTI3H9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273241H
Description
PANK2 Monoclonal specifically detects PANK2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PANK2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PANK2 | |
Full length recombinant protein of human PANK2 (NP_705902) produced in HEK293T cells. | |
0.1 mL | |
80025.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3H9 | |
Western Blot, Immunohistochemistry | |
C20orf48, EC 2.7.1.33, FLJ17232, Hallervorden-Spatz syndrome, HARPFLJ11729, hPanK2, HSS, MGC15053, NBIA1, pantothenate kinase 2, pantothenate kinase 2, mitochondrial, Pantothenic acid kinase 2, PKANneurodegeneration with brain iron accumulation 1 (Hallervorden-Spatz syndrome) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction