Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PANK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552563
Description
Immunogen sequence: SLHTVFCATG GGAYKFEQDF LTIGDLQLCK LDELDCLIKG ILYIDSVGFN GRSQCYYFEN PADSEKCQKL PFDLKNPYPL LLVNIGSGVS ILAVYSKDNY KRVTGT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Pantothenate kinase is an essential regulatory enzyme in CoA biosynthesis, catalyzing the cytosolic phosphorylation of pantothenate (vitamin B5), N-pantothenoylcysteine, and pantetheine. CoA is the major acyl carrier, playing a central role in intermediary and fatty acid metabolism. In both yeast and fly, each with only 1 pantothenate kinase gene, the null mutant is inviable. Mutations in PANK2 are the cause of pantothenate kinase-associated neurodegeneration (PKAN), formerly known as Hallervorden-Spatz syndrome (HSS). PKAN is an autosomal recessive neurodegenerative disorder associated with iron accumulation in the brain. Mutations in PANK2 are the cause of hypoprebetalipoproteinemia, acanthocytosis, retinitis pigmentosa, and pallidal degeneration (HARP).
Specifications
PANK2 | |
Polyclonal | |
Unconjugated | |
PANK2 | |
4933409I19Rik; AI642621; C20orf48; Hallervorden-Spatz syndrome; HARP; hPanK2; HSS; NBIA1; PANK2; pantothenate kinase 2; pantothenate kinase 2 (Hallervorden-Spatz syndrome); pantothenate kinase 2, mitochondrial; pantothenate kinase 2, mitochondrial isoform 1 preproprotein; pantothenate kinase 2, mitochondrial-like; pantothenic acid kinase 2; PKAN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80025 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZ23 | |
PANK2 | |
Recombinant protein corresponding to Human PANK2. Recombinant protein control fragment (Product #RP-89756). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction