Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PANK3 Antibody (OTI3C4), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273244MFV610
Description
PANK3 Monoclonal antibody specifically detects PANK3 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
PANK3 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
79646 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI3C4 | |
Flow Cytometry | |
DKFZp686O22247, EC 2.7.1.33, FLJ12899, hPanK3, MGC16863, pantothenate kinase 3, Pantothenic acid kinase 3 | |
Full length human recombinant protein of human PANK3 (NP_078870) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction