Learn More
Invitrogen™ PANK4 Monoclonal Antibody (4F10)

Description
Peptide Sequence: AGMDPVVHSA LQEERLLLVQ TGSSSPCLDL SRLDKGLAAL VRERGADLVV IEGMGRAVHT NYHAALRCES LKLAVIKNAW LAERLGGRLF SVIFKYEVPA E.
Specifications
Specifications
| Antigen | PANK4 |
| Applications | ELISA, Immunohistochemistry (PFA fixed) |
| Classification | Monoclonal |
| Clone | 4F10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Pank4 |
| Gene Accession No. | Q9NVE7 |
| Gene Alias | 4'-phosphopantetheine phosphatase; D030031I12Rik; DKFZp547M242; Fang1; FLJ10782; hPanK4; hypothetical protein Fang1; mPanK4; Pank4; Pantothenate kinase; Pantothenate kinase 4; pantothenic acid kinase 4; R75150; rPanK4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.