Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PANK4, p Thr63 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260782
Description
PANK4 Polyclonal specifically detects PANK4 in Mouse, Rat, Hamster samples. It is validated for Western Blot.Specifications
PANK4 (p Thr63) | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
DKFZp547M242, EC 2.7.1.33, FLJ10782, hPanK4, pantothenate kinase 4, Pantothenic acid kinase 4 | |
Rabbit | |
86 kDa | |
100 μL | |
Primary | |
Mouse, Rat, Hamster | |
IgG |
Western Blot | |
Unconjugated | |
10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
PANK4 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr63 conjugated to KLH. Accession # Q80YV4 | |
Antigen Affinity-purified | |
Cellular Signaling | |
55229 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction