Learn More
Invitrogen™ PANK4 Polyclonal Antibody

Description
Immunogen sequence: FDHSGKDTERE HEPPYEISVQ EEITARLHFI KFENTYIEAC LDFIKDHLVN TETKVIQATG GGAYKFKDLI EEKLRLKVDK EDVMTCLIKG CNFVLKNIPH EAFVYQKDSD PEFRFQTNHP HIFPYLLVNI GSGVSIVKVE TE Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Specifications
Specifications
| Antigen | PANK4 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Pank4 |
| Gene Accession No. | Q9NVE7 |
| Gene Alias | 4'-phosphopantetheine phosphatase; D030031I12Rik; DKFZp547M242; Fang1; FLJ10782; hPanK4; hypothetical protein Fang1; mPanK4; Pank4; Pantothenate kinase; Pantothenate kinase 4; pantothenic acid kinase 4; R75150; rPanK4 |
| Gene Symbols | Pank4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.