Learn More
Invitrogen™ PANK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149978

Description
PANK4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.
Specifications
PANK4 | |
Polyclonal | |
Unconjugated | |
Pank4 | |
4'-phosphopantetheine phosphatase; D030031I12Rik; DKFZp547M242; Fang1; FLJ10782; hPanK4; hypothetical protein Fang1; mPanK4; Pank4; Pantothenate kinase; Pantothenate kinase 4; pantothenic acid kinase 4; R75150; rPanK4 | |
Rabbit | |
Protein A | |
RUO | |
171053, 269614, 55229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q80YV4, Q923S8, Q9NVE7 | |
Pank4 | |
Recombinant Pantothenate Kinase 4 (PANK4) (Glu639-Glu773). | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.