Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PANK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552896 Supplier Invitrogen™ Supplier No. PA552896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDHSGKDTER EHEPPYEISV QEEITARLHF IKFENTYIEA CLDFIKDHLV NTETKVIQAT GGGAYKFKDL IEEKLRLKVD KEDVMTCLIK GCNFVLKNIP HEAFVYQKDS DPEFRFQTNH PHIFPYLLVN IGSGVSIVKV ETE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PANK4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pank4
Gene Accession No. Q9NVE7
Gene Alias 4'-phosphopantetheine phosphatase; D030031I12Rik; DKFZp547M242; Fang1; FLJ10782; hPanK4; hypothetical protein Fang1; mPanK4; Pank4; Pantothenate kinase; Pantothenate kinase 4; pantothenic acid kinase 4; R75150; rPanK4
Gene Symbols Pank4
Host Species Rabbit
Immunogen Recombinant protein fragment of Human PANK4 (Product #RP-88850)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55229
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.