Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pannexin-1 Antibody (28G5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Pannexin-1 |
---|---|
Clone | 28G5 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
Pannexin-1 Monoclonal antibody specifically detects Pannexin-1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
Pannexin-1 | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGC21309, MRS1innexin, pannexin 1, pannexin-1, PX1, UNQ2529 | |
A synthesized peptide derived from Human Pannexin-1 [UniProt Q96RD7] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
28G5 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
24145 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title