Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pannexin 1 Recombinant Rabbit Monoclonal Antibody (28G5)
GREENER_CHOICE

Catalog No. FEMA550140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550140 Supplier Invitrogen™ Supplier No. MA550140
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Pannexin 1 Recombinant Monoclonal Antibody for ELISA, Flow, ICC/IF, IHC (P)

The pannexin gene family encodes a second class of putative gap junction proteins and are highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pannexin 1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 28G5
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4
Gene PANX1
Gene Accession No. Q96RD7
Gene Alias AI847747; innexin; MRS1; Pannexin 1; pannexin-1; Panx1; px1; UNQ2529; UNQ2529/PRO6028
Gene Symbols PANX1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PANX1 (aa 366-426).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24145
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.