Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pannexin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542246 Supplier Invitrogen™ Supplier No. PA542246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IISELDKSYN RSIRLVQHML KIRQKSSDPY VFWNELEKAR KERYFEFPLL Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Pannexins constitute a new family of gap junction type proteins. They are highly conserved in worms, mollusks, insects and mammals and are predicted to have four transmembrane regions, two extracellular loops, one intracellular loop and intracellular N- and C-termini. Besides the connexin proteins, Pannexin 1 and 2 seem to be molecular components of neuronal gap junctions (electrical synapses) that are thought to form connections between principal cells in the hippocampus. Functional expression in paired Xenopus oocytes indicated that pannexins are capable of forming communicating junctions. In addition to forming gap junction channels in paired oocytes, Pannexin 1 can also form a mechanosensitive and ATP-permeable channel in the nonjunctional plasma membrane. Pannexin 1 and 3 are both glycoproteins with a cell surface distribution profile and life cycle dynamics distinct from connexin 43. Pannexin 3 expression was prevalent in skin and cartilage. In transiently expressing cells both pannexin 1 and 3 were incapable of forming intercellular channels but assembled into functional cell surface channels. Suggested roles in the ischemic death of neurons, schizophrenia, inflammation and tumor suppression have drawn attention for properties and cellular functions of pannexins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pannexin 3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PANX3
Gene Accession No. Q96QZ0
Gene Alias 3230401P04; 4833413G11Rik; gap junction protein pannexin 3; pannexin 3; pannexin-3; Panx3; Px3; si:dkey-263o8.2
Gene Symbols PANX3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PANX3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116337
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.