Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Panobinostat

Catalog No. AC467000010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467000010 1 g
AC467002500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467000010 Supplier Thermo Scientific Chemicals Supplier No. 467000010
Add to Cart

CAS: 404950-80-7 | C21H23N3O2, C21H23N3O2 | 349.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404950-80-7
Molecular Formula C21H23N3O2

Specifications

Quantity 1 g
Formula Weight 349.43
InChI Key FPOHNWQLNRZRFC-UHFFFAOYSA-N
SMILES CC1=C(CCNCC2=CC=C(C=CC(=O)NO)C=C2)C2=CC=CC=C2N1
IUPAC Name N-hydroxy-3-[4-({[2-(2-methyl-1H-indol-3-yl)ethyl]amino}methyl)phenyl]prop-2-enamide
Molecular Weight (g/mol) 349.43
Chemical Name or Material Panobinostat
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Toxic if inhaled.

Causes skin irritation.

Causes serious eye damage.

May damage fertility or the unborn child.

May cause damage to organs through prolonged or repeated exposure.<br/
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.