Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PANP/PILR alpha associated neural protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$377.03 - $665.78
Specifications
| Antigen | PANP/PILR alpha associated neural protein |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PANP/PILR alpha associated neural protein Polyclonal specifically detects PANP/PILR alpha associated neural protein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PANP/PILR alpha associated neural protein | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 196500 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DRSQKRRRPSGQQGALRQEESQQPLTDLSPAGVTVLGAFGDSPTPTPDHEEPRGGPRPGMPHPKGAPAFQLNR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C12orf53, chromosome 12 open reading frame 53, PANP, PIANP, PILR alpha associated neural protein | |
| PIANP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title