Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PANP/PILR alpha associated neural protein Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2833540.1ML
Description
PANP/PILR alpha associated neural protein Polyclonal specifically detects PANP/PILR alpha associated neural protein in Human samples. It is validated for Western Blot.Specifications
PANP/PILR alpha associated neural protein | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
196500 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
C12orf53, chromosome 12 open reading frame 53, PANP, PIANP, PILR alpha associated neural protein | |
The immunogen is a synthetic peptide directed towards the middle region of human PANP/PILR alpha associated neural protein | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction