Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PANX1 Antibody, ProSci
SDP

Catalog No. 16830309
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-830-309 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-830-309 Supplier ProSci LLC Supplier No. 298000100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PANX1 Polyclonal Antibody specifically detects PANX1 in Human samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Specifications

Antigen PANX1
Applications ELISA, Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution PANX1 antibody can be used for detection of PANX1 by ELISA at 1:62500. PANX1 antibody can be used for detection of PANX1 by western blot at 1.0 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Formulation Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Gene Accession No. NP_056183
Gene Alias PANX1, MGC21309, MRS1, UNQ2529, PX1
Host Species Rabbit
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human PANX1.
Purification Method Antibody is purified by peptide affinity chromatography method.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24145
Target Species Human
Content And Storage For short periods of storage (days) store at 4°C. For longer periods of storage, store PANX1 antibody at -20°C. As with any antibody avoid repeat freeze-thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Membrane , Inflammasomes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.