Learn More
Invitrogen™ PAOX Polyclonal Antibody

Description
PAOX Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PAOX |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Paox |
| Gene Accession No. | Q6QHF9, Q8C0L6 |
| Gene Alias | 2410012F02Rik; AI118225; exo-N4-amino; PAO; Paox; Peroxisomal N(1)-acetyl-spermine/spermidine oxidase; peroxisomal N1-acetyl-spermine/spermidine oxidase; polyamine oxidase; polyamine oxidase (exo-N4-amino); polyamine oxidase (exo-N4-amino) isoform 1; putative GTP-binding protein; UNQ1923/PRO4398 |
| Gene Symbols | Paox |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.