Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Papain (Papaya), 2X

Catalog No. 89955674
Click to view available options
Quantity:
25 mg

Used for Enzyme

#REF!

Specifications

Accession Number P00784
For Use With (Application) Enzyme Activity
Formulation In 50mM sodium acetate, pH 4.5
Molecular Weight (g/mol) 23.4kDa
Name Papain (Papaya), 2x
Preparation Method Native protein purified from Papaya Latex
Quantity 25 mg
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name PAPAIN
Assays Papain Assay; Cell isolation (more gentle than other proteases); Protein structural studies; Peptide mapping; Red cell surface modification for antibody screening or identification; Fab preparation from IgG and IgM antibodies; Solubilization of integral membrane proteins; Production of glycopeptides from purified proteoglycans; Enzymatic wound debridement.
Biological Activity Activates to at least 20 units per mg protein. One Unit hydrolyzes one micromole of benzoyl-L-arginine ethyl ester per minute at 25°C, pH 6.2, after activation in a solution containing 1.1 mM EDTA, 0.067 mM mercaptoethanol and 5.5 mM cysteine-HCl for 30 minutes.
Species Plants
Protein Tag None
Expression System Native protein purified from Papaya Latex
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.