Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Papain (Papaya)

Catalog No. 89955672 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Tub

Used for Enzyme

#REF!

Specifications

Accession Number P00784
For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 23.4kDa
Name Papain (Papaya)
Preparation Method Native protein purified from Papaya Latex
Quantity 1 Tub
Storage Requirements Store at 4°C. Reconstituted with 5 ml of Earle's Balanced Salt Solution (EBSS) or equivalent yields a solution at 20 units of papain per ml in 1 mM L-cysteine with 0.5 mM EDTA. Do not freeze the reconstituted solution.
Regulatory Status RUO
Common Name PAPAIN
Assays Papain Assay; Cell isolation (more gentle than other proteases); Protein structural studies; Peptide mapping; Red cell surface modification for antibody screening or identification; Fab preparation from IgG and IgM antibodies; Solubilization of integral membrane proteins; Production of glycopeptides from purified proteoglycans; enzymatic wound debridement.
Biological Activity ≥100 units per vial. One Unit hydrolyzes one micromole of benzoyl-L-arginine ethyl ester per minute at 25°C, pH 6.2, after activation in a solution containing 1.1mM EDTA, 0.067 mM mercaptoethanol and 5.5mM cysteine-HCl for 30 minutes.
Species Plants
Protein Tag None
Expression System Native protein purified from Papaya Latex
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.