Learn More
Epredia™ Papanicolaou OG-6 and Modified OG Stains
Catalog No. p-4530219 Shop All Epredia Products
Click to view available options
Quantity:
1 pt.
1 gal.
2.5 gal.
Type:
Modified OG Stain
OG-6 Papanicolaou Stain
Stain gynecological specimens with Epredia™ Papanicolaou Stain OGs.
  • OG-6 stains the keratinized areas of the cell
  • Modified OG is a Gill modification that minimizes precipitation and reduces staining time
  • Use OG-6 with EA-36 or EA-50 when staining gynecological specimens
  • Use modified OG with EA-65 for staining non-gynecological specimens
Specifications
Type OG-6 Papanicolaou Stain
Quantity 1 pt.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.