Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papanicolaou Stain, EA-65 Formula, Ricca Chemical
SDP

Catalog No. 55051 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle

Counterstain for use with Hematoxylin Stains in Papanicolaou Staining

Specifications

Chemical Name or Material Papanicolaou Stain
Name Note EA-65 Formula
CAS 64-17-5, 7732-18-5, 67-56-1, 17372-87-1, 12501-23-4, 10114-58-6
CAS Min % 91.0, 6.07, 0.27, 0.24, 0.06, 0.02
CAS Max % 95.0, 6.32, 0.28, 0.25, 0.06, 0.02
Grade Laboratory
Packaging Natural Poly Bottle
Color Green
Quantity 4 L
Physical Form Murky Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.