Learn More
ENG Scientific Papanicolaou Staining Solutions
Catalog No. p-3274911
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
Type:
OG-6 Papanicolaou Stain
This item is not returnable. View return policy
This item is not returnable. View return policy
Smears are alcohol-ether fixed.
  • Solutions are made from certified dyes (where applicable)
  • Results: nuclei stain blue with clear, sharp detail; cytoplasm stain varying shades of pink, blue, yellow, green, and gray
  • Eosin Y, Bismark Brown Y, Light Green SF, Yellowish, Orange G, Ethanol, Phosphotungstic Acid
Specifications
Type OG-6 Papanicolaou Stain
For Use With (Application) For use on smears of bronchial aspirates, sputum, pleural and peritoneal fluids
Includes Eosin Y, bismark brown Y, light green SF, yellowish, orange G, ethanol, phosphotungstic acid
Packaging Bottle
Quantity 1 x 3800 mL (1 gal.)
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.