Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPD1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222258PCP
Description
PAPD1 Polyclonal antibody specifically detects PAPD1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| PAPD1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| EC 2.7.7.19, FLJ10486, mitochondrial poly(A) polymerase, mtPAP, PAP, PAP associated domain containing 1, PAP-associated domain-containing protein 1, PAPD1SPAX4, poly(A) RNA polymerase, mitochondrial, Polynucleotide adenylyltransferase, Terminal uridylyltransferase 1, TUTase 1 | |
| The immunogen this antibody was made to, maps to a region between residue 532 and 582 of human PAP-Associated Domain-Containing Protein 1 using the numbering given in entry NP_060579.3 (GeneID 55149). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55149 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction