Learn More
PAPD1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303904AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the DNA polymerase type-B-like family. This enzyme synthesizes the 3' poly(A) tail of mitochondrial transcripts and plays a role in replication-dependent histone mRNA degradation.Specifications
PAPD1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NVV4 | |
MTPAP | |
Between 532 and 582 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MTPAP | |
PAPD1, SPAX4, PAP-associated domain-containing protein 1, TUTase 1, poly(A) RNA polymerase, mitochondrial, polynucleotide adenylyltransferase, terminal uridylyltransferase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55149 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.