Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | PAPD4 |
---|---|
Dilution | Western Blot 1:1000, ELISA 1:100-1:1000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PAPD4 Polyclonal antibody specifically detects PAPD4 in Human samples. It is validated for Western Blot, ELISASpecifications
PAPD4 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
Q6PIY7 | |
167153 | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000, ELISA 1:100-1:1000 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antiserum | |
EC 2.7.7.19, FLJ38499, GLD2TUTase 2, hGLD-2, PAP associated domain containing 4, PAP-associated domain-containing protein 4, poly(A) RNA polymerase GLD2, Terminal uridylyltransferase 2 | |
Polyclonal antibody raised in rabbit against human PAPD4 (PAP associated domain containing 4), using a KLH-conjugated synthetic peptide containing a sequence from the N-terminal part of the protein. | |
Whole Antiserum |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title