Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Paper Chromatography Kit
GREENER_CHOICESCI_ED

Catalog No. s96896 Shop All Aldon Products
Click to view available options
Class Size:
15 Lab Groups
WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Includes

Materials for 15 groups: Chromatography dye set with 0.5mL each of Crystal Violet, Safranin O, Toluidine Blue and a chromatography mixture; 30 mL 95% Ethanol UN1170 (5); chromatography sheets (15); and capillary tubes (4)

Requires

Plastic wrap or tinfoil, metric rulers, distilled water, 250 mL beakers (15) and 25 mL graduated cylinder

Investigate the oldest documented technique used to separate chemical substances.

  • Perform paper chromatography on three individual dyes and a mixture of dyes Determine if all three dyes are in the mixture
  • See why different substances move at different rates within the chromatography system

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Investigate The Oldest Documented Technique Used to Separate Chemical Substances
Includes Chromatography dye set with 0.5 mL crystal violet, 0.5 mL safranin o, 0.5 mL toluidine blue, 0.5 mL chromatography mixture, 5 x 30 mL 95% ethanol, 15 chromatography sheets, 4 capillary tubes
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Paper Chromatography Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.