Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDC Healthcare™ Paper with Permanent Adhesive
Click to view available options
Dimensions (LxW):
1 x 3 in.
2 x 2 in.
2.35 x 0.87 in.
2.5 x 1.25 in.
3 x 1.62 in.
Description
- Pre-printed with 0.1% FORMALIN text for consistent hazard identification
- Permanent adhesive ensures labels remain firmly attached to containers
- Fluorescent pink coloring provides excellent visibility in laboratory settings
- Paper construction allows for clear printing and durability
- Roll format with 1,000 labels enables efficient application
Specifications
Specifications
Adhesive Type | Permanent |
Coating | None |
Color | White |
Finish | Matte |
Format | Roll |
Material | Paper |
Product Type | Labels |
Sterility | Non-sterile |
Shape | Diamond |
Dimensions (LxW) | 2 x 2 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction