Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Papilin Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305954PE
Description
Papilin Polyclonal antibody specifically detects Papilin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Papilin | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
papilin, Papilin, Proteoglycan-Like Sulfated Glycoprotein, PAPLN | |
Produced in rabbits immunized with E. coli-derived Human Papilin fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
89932 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction