Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAPL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572659
Description
PAPL Polyclonal Antibody for Western Blot

Specifications
PAPL | |
Polyclonal | |
Unconjugated | |
ACP7 | |
acid phosphatase 7, tartrate resistant (putative); acid phosphatase type 7; acp7; iron/zinc purple acid phosphatase-like protein; PAPL; PAPL1; Purple acid phosphatase long form; purple acid phosphatase long form 1; zgc:162913 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
571830 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.67 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
A5D6U8 | |
ACP7 | |
KLH conjugated synthetic peptide between 378-413 amino acids of DANRE papl. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction