Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAPLN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565791
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565791 Supplier Invitrogen™ Supplier No. PA565791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACSLEDRPPLT EPCVHEDCPL LSDQAWHVGT WGLCSKSCSS GTRRRQVICA IGPPSHCGSL QHSKPVDVEP CNTQPCHLPQ EVPSMQDVHT PA Highest antigen sequence identity to the following orthologs - mouse 67%, rat 69%.

PAPLN (Papilin, Proteoglycan Like Sulfated Glycoprotein) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include peptidase activity and serine-type endopeptidase inhibitor activity. An important paralog of this gene is ADAMTS9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPLN
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAPLN
Gene Accession No. O95428
Gene Alias E030033C16Rik; Papilin; papilin, proteoglycan like sulfated glycoprotein; papilin, proteoglycan-like sulfated glycoprotein; Papln; UNQ2420/PRO4977
Gene Symbols PAPLN
Host Species Rabbit
Immunogen Recombinant Human PAPLN. Recombinant protein control fragment (Product #RP-104968).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89932
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.