Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPOG Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS4968R

Description
This gene overlaps with the gene for cysteineendopeptidase AUT-like 4 in a head-to-tail orientation.Specifications
PAPOG | |
Polyclonal | |
Unconjugated | |
PAPOLG | |
9630006B20Rik; BOS_11496; FLJ11805; FLJ12972; FLJ13482; FLJ14187; hypothetical protein MGC63762; MGC133307; MGC133308; Neo-PAP; Neo-poly(A) polymerase; nuclear poly(A) polymerase gamma; PAP2; PAPG; PAP-gamma; PAPOLG; Poly(A) polymerase gamma; Polynucleotide adenylyltransferase gamma; Signal recognition particle RNA-adenylating enzyme; SRP 3′-adenylating protein; SRP RNA 3′ adenylating enzyme/pap2; SRP RNA 3′-adenylating enzyme; SRP RNA-adenylating enzyme; zgc:63762 | |
Rabbit | |
Protein A | |
RUO | |
64895 | |
-20°C | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9BWT3 | |
PAPOLG | |
KLH conjugated synthetic peptide derived from human PAPOG. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction