Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPOLA Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055267C
Description
PAPOLA Polyclonal antibody specifically detects PAPOLA in Human samples. It is validated for Western BlotSpecifications
PAPOLA | |
Polyclonal | |
Western Blot | |
EC 2.7.7, EC 2.7.7.19, PAP-alpha, PAPMGC5378, poly(A) polymerase alpha, Polynucleotide adenylyltransferase alpha | |
The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human poly(A) polymerase alpha using the numbering given in entry NP_116021.2 (GeneID 10914). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10914 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction