Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAPOLA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551634 Supplier Invitrogen™ Supplier No. PA551634
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGVKLTALND SSLDLSMDSD NSMSVPSPTS ATKTSPLNSS GSSQGRNSPA PAVTAASVTN IQATEVSVPQ VNSSESSGGT SSESIPQTAT QPAISPPPKP TVSRVVSSTR LVNPPPRSSG NAATSGNAAT KIPTPIVGVK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Polymerase that creates the 3'-poly(A) tail of mRNA's. Also required for the endoribonucleolytic cleavage reaction at some polyadenylation sites. May acquire specificity through interaction with a cleavage and polyadenylation specificity factor (CPSF) at its C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPOLA
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAPOLA
Gene Accession No. P51003
Gene Alias Pap; PAP-alpha; PapIII; PAPOLA; Plap; poly (A) polymerase alpha; poly(A) polymerase alpha; Polynucleotide adenylyltransferase; polynucleotide adenylyltransferase alpha
Gene Symbols PAPOLA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAPOLA. Recombinant protein control fragment (Product #RP-88993).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10914
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.