Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAPOLB Monoclonal Antibody (3D7)
GREENER_CHOICE

Catalog No. PIMA522298
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522298 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522298 Supplier Invitrogen™ Supplier No. MA522298
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MMPFPVTTQG PPQPAPPPNR YGVSSPISLA VPKETDCLLT QRLIETLRPF GVFEEEEELQ RRILVLDKLN NLVKEWIREI SESKSLPQSV IENVGGKIFT FGSYRLGVHT KGADIDALCV APSHVDRSDF FTSFYAKLKL QEEVKDLRAV QEAFVPVIKL CFDGIEIDIL FARLALQTIP EDLDLRDDSL LKNLDIRCIR SLNGCRVTDE ILHLVPNIDN FRLTLRAIKL WAKCHNIYSN ILGFLGGVSW AMLVARTCQL YPNAVASTLV RKFFLVFSEW EWPNPVLLKE PEERNLNLPV WDPRVNPSDR YHLMPIITPA YPQQNSTYNV SISTRMVMIE EFKQGLAITH EILLSKAEWS KLFEAPSFFQ KYKHYIVLLA SASTEKQHLE WVGLVESKIR ILVGSLEKNE FITLAHVNPQ SFPAPKENPD MEEFRTMWVI GLGLKKPDNS EILSIDLTYD IQSFTDTVYR QAVNSKMFEM GMKITAVHLR RKELHQLLPH HVLQDKKAHS TEGRRLTDLN DSSFDLSAGC ENSMSVPSST STMKTGPLIS SSQGRNSPAL AVMTASVANI QATEFSLQQV NTNESSGVAL NESIPHAVSQ PAISPSPKAM VARVVSSTCL ISHPDLQETQ QQTYLIL.

Testis-specific poly(A) polymerase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPOLB
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3D7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PAPOLB
Gene Accession No. Q9NRJ5
Gene Alias Med21; PAP; PAP-beta; Paplob; Papola-ps; PAPOLB; PAPT; Plap-ps; poly (A) polymerase beta (testis specific); poly(A) polymerase beta; poly(A) polymerase beta (testis specific); polynucleotide adenylyltransferase beta; Testis-specific poly(A) polymerase; TPAP
Gene Symbols PAPOLB
Host Species Mouse
Immunogen PAPOLB (AAH36653, 1 a.a. ~ 637 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56903
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.