Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPOLB Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | PAPOLB |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PAPOLB Polyclonal antibody specifically detects PAPOLB in Human samples. It is validated for ImmunofluorescenceSpecifications
PAPOLB | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
EC 2.7.7, EC 2.7.7.19, PAP-beta, PAPTTPAP, poly(A) polymerase beta, poly(A) polymerase beta (testis specific), Polynucleotide adenylyltransferase beta, Testis-specific poly(A) polymerase | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SLQQVNTNESSGVALNESIPHAVSQPAISPSPKAMVARVVSSTCLISHPDLQETQQQTYLIL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
56903 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title